Anti-ARRS Mouse Monoclonal Antibody [clone: S128]
Lieferant: Biosensis
M-1796-100
BSENM-1796-100EA 312 EUR
BSENM-1796-100
Anti-ARRS Mouse Monoclonal Antibody [clone: S128]
Antikörper
Binds to photoactivated, phosphorylated RHO and terminates RHO signaling via G-proteins by competing with G-proteins for the same binding site on RHO (PubMed: 8003967, PubMed: 25205354). May play a role in preventing light-dependent degeneration of retinal photoreceptor cells (By similarity). Ref: uniprot.org. Antibody detects arrestin-1 protein only and does not cross-react with the other 3 arrestin molecules.
Western blotting (1:5,000) and Immunohistochemistry (1:1,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: ARRS
Clonality: Monoclonal
Clone: S128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Pig