Anti-MECP2 Mouse Monoclonal Antibody [clone: 4F11]
Lieferant: Biosensis
M-1809-100
BSENM-1809-100EA 312 EUR
BSENM-1809-100
Anti-MECP2 Mouse Monoclonal Antibody [clone: 4F11]
Antikörper
Chromosomal protein that binds to methylated DNA. It can bind specifically to a single methyl-CpG pair. It is not influenced by sequences flanking the methyl-CpGs. Mediates transcriptional repression through interaction with histone deacetylase and the corepressor SIN3A. Binds both 5-methylcytosine (5mC) and 5-hydroxymethylcytosine (5hmC)-containing DNA, with a preference for 5-methylcytosine (5mC). Ref: uniprot.org
Western blotting (1:5,000-1:10,000) and Immunohistochemistry (1:1,000-1:5,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MECP2
Clonality: Monoclonal
Clone: 4F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat