Anti-OPSD Mouse Monoclonal Antibody [clone: B630]

Lieferant: Biosensis
M-1815-100
BSENM-1815-100EA 312 EUR
BSENM-1815-100
Anti-OPSD Mouse Monoclonal Antibody [clone: B630]
Antikörper
Photoreceptor required for image-forming vision at low light intensity. Required for photoreceptor cell viability after birth (By similarity). Light-induced isomerization of 11-cis to all-trans retinal triggers a conformational change that activates signaling via G-proteins (PubMed:10926528, PubMed:12044163, PubMed:11972040, PubMed:16908857, PubMed:16586416, PubMed:17060607, PubMed:17449675, PubMed:18818650, PubMed:21389983, PubMed:22198838, PubMed:23579341, PubMed:25205354, PubMed:27458239). Subsequent receptor phosphorylation mediates displacement of the bound G-protein alpha subunit by the arrestin SAG and terminates signaling (PubMed:1396673, PubMed:15111114). Ref: uniprot.org

Western blotting (1:5,000) and Immunohistochemistry (1:1,000). Due to the highly hydrophobic nature of rhodopsin, avoid boiling samples in SDS-PAGE sample buffer for rhodopsin analysis by Western Blotting, as this will result in extensive aggregation of the rhodopsin protein and appearance of high molecular weight bands. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: OPSD
Clonality: Monoclonal
Clone: B630
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Cow, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR