TISSUE SPECIFICITY: Ubiquitously expressed. Expressed in dorsal root ganglia, trigeminal ganglia, spinal chord (Lissauer's tract, dorsal horn and dorsal columns) (at protein level). PTM: N-glycosylated. PTM: Phosphorylated by PKA. SIMILARITY: Belongs to the transient receptor family. TrpV subfamily. SIMILARITY: Contains 3 ANK repeats.
Immunohistochemical analysis in rat dorsal root ganglia and spinal cord indicates a high level of specificity for this antiserum. Specificity was also shown by Western blot.
Application Information:
IHC, Immunofluorescence, Western blot and Immunoblot. Recommended to be used at a concentration of 0.5-1 µg for these applications. This antiserum works superbly for immunohistochemistry on free-floating or paraffin embedded sections. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: VRL-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: at VRL-1 protein (aa: 744-761)
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human