Anti-His Tag Rabbit Polyclonal Antibody

Lieferant: Biosensis
R-1312-100
BSENR-1312-100EA 388 EUR
BSENR-1312-100
Anti-His Tag Rabbit Polyclonal Antibody
Antikörper
The 6X His tag is a short peptide sequence of 6 histidine residues. Epitopes such as the 6X His tag are often included with the target DNA at the time of cloning to produce fusion proteins containing the tag sequence. This allows anti-epitope tag antibodies such as this one to serve as a universal detection reagent for any recombinant protein containing this tag. Anti-epitope antibodies are a useful alternative to generating antibodies to identify a specific recombinant protein. The 6X His motif is often used as a tag on recombinant proteins to facilitate purification with immobilized metal-affinity chromatography.

His-tagged fusion proteins
Application Information:
ELISA, WB, IP, IF, IHC. Western Blotting at a suggested concentration of 1:500~1:1000. ELISA at a suggested concentration of 1: 2500. IP 5 µg/sample. IF 5 µg/mL. IHC 5 µg/mL. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Tag
Antigen: His Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 6-his peptide
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR