Anti-MMP2 Rabbit Polyclonal Antibody

Lieferant: Biosensis
R-1381-100
BSENR-1381-100EA 312 EUR
BSENR-1381-100
Anti-MMP2 Rabbit Polyclonal Antibody
Antikörper
Ubiquitinous metalloproteinase that is involved in diverse functions such as remodeling of the vasculature, angiogenesis, tissue repair, tumor invasion, inflammation, and atherosclerotic plaque rupture. As well as degrading extracellular matrix proteins, can also act on several nonmatrix proteins such as big endothelial 1 and beta-type CGRP promoting vasoconstriction. Also cleaves KISS at a Gly-|-Leu bond. Appears to have a role in myocardial cell death pathways. Contributes to myocardial oxidative stress by regulating the activity of GSK3beta. Cleaves GSK3beta in vitro. Involved in the formation of the fibrovascular tissues in association with MMP14. PEX, the C-terminal non-catalytic fragment of MMP2, posseses anti-angiogenic and anti-tumor properties and inhibits cell migration and cell adhesion to FGF2 and vitronectin. Ligand for integrinv/beta3 on the surface of blood vessels. MMP2 isoform 2 mediates the proteolysis of CHUK/IKKA and initiates a primary innate immune response by inducing mitochondrial-nuclear stress signaling with activation of the pro-inflammatory NF-kappaB, NFAT and IRF transcriptional pathways. Catalytic activity of MMP2 causes cleavage of gelatin type I and collagen types IV, V, VII, X. Cleaves the collagen-like sequence Pro-Gln-Gly-|-Ile-Ala-Gly-Gln. (Ref: uniprot.org).

Western Blotting (WB): 1:5,000 - 1:10,000. MMP2 appears as two bands at apparent molecular weights of 66 and 72 kDa.

Immunohistochemistry (IHC): 1:500-1:1,000.

Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human C-terminal peptide (amino acids 475-490).
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR