Anti-Beta Endorphin Rabbit Polyclonal Antibody

Lieferant: Biosensis
R-1492-50
BSENR-1492-50EA 321 EUR
BSENR-1492-50
Anti-Beta Endorphin Rabbit Polyclonal Antibody
Antikörper
Human beta-endorphin is a 31 amino acid peptide cleaved from the precursor pro-opiomelanocortin (POMC). It is an endogenous opioid peptide neurotransmitter that interacts with opioid receptors.

Confirmed by RIA against the synthetic peptide.
Application Information:
A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. For radioimmunoassay (RIA), a working dilution of 1:10,000 is recommended. Sensitivity - 120 pg/sample; IC50-1 ng/ sample. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Beta Endorphin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple epitopes because of the nature of the immunogen
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR