Anti-pan synuclein Sheep Polyclonal Antibody

Lieferant: Biosensis
S-044-100
BSENS-044-100EA 321 EUR
BSENS-044-100
Anti-pan synuclein Sheep Polyclonal Antibody
Antikörper
NULL

Overlap specific immunohistochemical staining of alpha-, beta- and gamma synucleins
Application Information:
IHC, WB. A concentration of 3 µg/ml is recommended for immunohistochemistry (frozen & paraffin embedded sections) and 1 µg/ml for western blot. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: pan synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: aa 11-22 Human alpha-, beta-, and gamma synuclein proteins
Host: Sheep
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR