Anti-DP-MHC class II Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BM-1]
Lieferant: Biotium
BNUM0061-50
BTIUBNUM0061-50EA 417 EUR
BTIUBNUM0061-50
Anti-DP-MHC class II Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BM-1]
Antikörper
Recognizes 183 kDa protein with DNA-binding characteristics, which is identified as a myeloid specific antigen. It reacts with myeloid precursor cells and granulocytes in bone marrow. Its antigen appears to be restricted to M2 and M3 acute myelogenous leukemia (AML) subtypes. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. This MAb reacts with early precursor and mature forms of human myeloid cells. It is useful in the identification of myelogenous leukemias, distinguishing granulocytic sarcomas from lymphoid malignancies and also in the study of differentiation and transformation of human myeloid cells. The biological function of this antigen is not clear, although it has been proposed that it may play a role in the differentiation of myeloid cells.
Type: Primary
Antigen: DP-MHC class II
Clonality: Monoclonal
Clone: BM-1
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human