Anti-SHBG Mouse Monoclonal Antibody (Purified, BSA-free) [clone: SHBG/245]

Lieferant: Biotium
BNUM0245-50
BTIUBNUM0245-50EA 417 EUR
BTIUBNUM0245-50
Anti-SHBG Mouse Monoclonal Antibody (Purified, BSA-free) [clone: SHBG/245]
Antikörper
Recognizes a protein of 45 kDa, identified as SHBG. It functions as an androgen transport protein, but may also be involved in receptor-mediated processes. Each dimer binds one molecule of steroid. It is specific for 5-alpha-dihydrotestosterone, testosterone, and 17-beta-estradiol. SHBG regulates the plasma metabolic clearance rate of steroid hormones by controlling their plasma concentration. In testis, it is synthesized by the Sertoli cells, secreted into the lumen of the seminiferous tubule and transported to the epididymis.

Type: Primary
Antigen: SHBG
Clonality: Monoclonal
Clone: SHBG/245
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR