Anti-ESR1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ESR1/420]
Lieferant: Biotium
BNUM0420-50
BTIUBNUM0420-50EA 417 EUR
BTIUBNUM0420-50
Anti-ESR1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ESR1/420]
Antikörper
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa300-550. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.
Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ESR1/420
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human