Anti-ESR1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ER506]

Lieferant: Biotium
BNUM0506-50
BTIUBNUM0506-50EA 417 EUR
BTIUBNUM0506-50
Anti-ESR1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ER506]
Antikörper
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. It strongly stains nuclei of epithelial cells in breast carcinomas.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ER506
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR