Anti-Tubulin beta Rabbit Polyclonal Antibody
Lieferant: Bioworld Technology
AP0064
BWRLAP0064EA 246 EUR
BWRLAP0064
Anti-Tubulin beta Rabbit Polyclonal Antibody
Antikörper
Recombinant full length Human Tubulin beta.
Tubulin beta 1/2/3/5/6/8 pAb pAb detects endogenous levels of Tubulin beta 1/2/3/5/6/8 protein.
Type: Primary
Antigen: Tubulin beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat