Anti-Smad2/3 Rabbit Polyclonal Antibody [clone: N175]

Lieferant: Bioworld Technology
AP0445
BWRLAP0445EA 262 EUR
BWRLAP0445
Anti-Smad2/3 Rabbit Polyclonal Antibody [clone: N175]
Antikörper
Synthetic peptide, corresponding to amino acids 145-195 of Human Smad2.

Smad2/3 (N175) pAb detects endogenous levels of Smad2/3 protein.

Type: Primary
Antigen: Smad2/3
Clonality: Polyclonal
Clone: N175
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR