Anti-SSB Rabbit Polyclonal Antibody [clone: T362]
Lieferant: Bioworld Technology
AP0467
BWRLAP0467EA 262 EUR
BWRLAP0467
Anti-SSB Rabbit Polyclonal Antibody [clone: T362]
Antikörper
Synthetic peptide, corresponding to amino acids 331-380 of Human SSB.
SSB(T362) pAb detects endogenous levels of SSB protein.
Type: Primary
Antigen: SSB
Clonality: Polyclonal
Clone: T362
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat