Anti-MRP8 Rabbit Polyclonal Antibody

Lieferant: Bioworld Technology
AP0476
BWRLAP0476EA 262 EUR
BWRLAP0476
Anti-MRP8 Rabbit Polyclonal Antibody
Antikörper
Synthetic peptide, corresponding to amino acids CMGVAAHKKSHEESHKE of Human MRP8.

MRP8 pAb detects endogenous levels of MRP8 protein.

Type: Primary
Antigen: MRP8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR