Anti-p-ERK1/2 Rabbit Polyclonal Antibody [clone: Y204]

Lieferant: Bioworld Technology
AP0490
BWRLAP0490EA 284 EUR
BWRLAP0490
Anti-p-ERK1/2 Rabbit Polyclonal Antibody [clone: Y204]
Antikörper
Synthetic phosphopeptide derived from human ERK1/2 around the phosphorylation site of Tyrosine 204.

p-ERK1/2 (Y204) pAb detects endogenous levels of ERK1/2 protein only when phosphorylated at tyrosine 204 protein.

Type: Primary
Antigen: p-ERK1/2
Clonality: Polyclonal
Clone: Y204
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR