Anti-TRAF2 Rabbit Polyclonal Antibody [clone: S11]

Lieferant: Bioworld Technology
AP0694
BWRLAP0694EA 262 EUR
BWRLAP0694
Anti-TRAF2 Rabbit Polyclonal Antibody [clone: S11]
Antikörper
Synthetic peptide, corresponding to the N-terminus of Human TRAF2.

TRAF2(S11) pAb detects endogenous levels of TRAF2 protein.

Type: Primary
Antigen: TRAF2
Clonality: Polyclonal
Clone: S11
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR