Anti-14-3-3 zeta Rabbit Polyclonal Antibody [clone: V52]

Lieferant: Bioworld Technology
BS1001
BWRLBS1001EA 262 EUR
BWRLBS1001
Anti-14-3-3 zeta Rabbit Polyclonal Antibody [clone: V52]
Antikörper
Synthetic peptide, corresponding to amino acids 20-70 of Human 14-3-3 zeta.

14-3-3 ? (V52) pAb detects endogenous levels of 14-3-3 protein zeta/delta.

Type: Primary
Antigen: 14-3-3 zeta
Clonality: Polyclonal
Clone: V52
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR