Anti-Chk1 Rabbit Polyclonal Antibody [clone: K274]

Lieferant: Bioworld Technology
BS1051
BWRLBS1051EA 262 EUR
BWRLBS1051
Anti-Chk1 Rabbit Polyclonal Antibody [clone: K274]
Antikörper
Synthetic peptide, corresponding to amino acids 251-300 of Human Chk1.

Chk1 (K274) pAb detects endogenous levels of Chk1 protein.

Type: Primary
Antigen: Chk1
Clonality: Polyclonal
Clone: K274
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR