Anti-CBP Rabbit Polyclonal Antibody [clone: V2427]

Lieferant: Bioworld Technology
BS1078
BWRLBS1078EA 262 EUR
BWRLBS1078
Anti-CBP Rabbit Polyclonal Antibody [clone: V2427]
Antikörper
Synthetic peptide, corresponding to amino acids 2395-2440 of Human CBP.

CBP (V2427) pAb detects endogenous levels of CBP protein.

Type: Primary
Antigen: CBP
Clonality: Polyclonal
Clone: V2427
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR