Anti-Cyclin A1 Rabbit Polyclonal Antibody [clone: R436]
Lieferant: Bioworld Technology
BS1084
BWRLBS1084EA 262 EUR
BWRLBS1084
Anti-Cyclin A1 Rabbit Polyclonal Antibody [clone: R436]
Antikörper
Synthetic peptide, corresponding to amino acids 400-450 of Human Cyclin A1.
Cyclin A1 (R436) pAb detects endogenous levels of Cyclin A1 protein.
Type: Primary
Antigen: Cyclin A1
Clonality: Polyclonal
Clone: R436
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat