Anti-Cytochrome c Rabbit Polyclonal Antibody [clone: H19]

Lieferant: Bioworld Technology
BS1089
BWRLBS1089EA 262 EUR
BWRLBS1089
Anti-Cytochrome c Rabbit Polyclonal Antibody [clone: H19]
Antikörper
Synthetic peptide, corresponding to the N-terminal of Human Cytochrome c.

Cytochrome c (H19) pAb detects endogenous levels of Cytochrome c protein.

Type: Primary
Antigen: Cytochrome c
Clonality: Polyclonal
Clone: H19
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR