Anti-EEF2 Rabbit Polyclonal Antibody [clone: A50]

Lieferant: Bioworld Technology
BS1099
BWRLBS1099EA 262 EUR
BWRLBS1099
Anti-EEF2 Rabbit Polyclonal Antibody [clone: A50]
Antikörper
Synthetic peptide, corresponding to amino acids 20-60 of Human eEF2.

eEF2 (A50) pAb detects endogenous levels of eEF2 protein.

Type: Primary
Antigen: EEF2
Clonality: Polyclonal
Clone: A50
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR