Anti-G3BP-1 Rabbit Polyclonal Antibody [clone: D226]

Lieferant: Bioworld Technology
BS1132
BWRLBS1132EA 262 EUR
BWRLBS1132
Anti-G3BP-1 Rabbit Polyclonal Antibody [clone: D226]
Antikörper
Synthetic peptide, corresponding to amino acids 196-246 of Human G3BP-1.

G3BP-1 (D226) pAb detects endogenous levels of G3BP-1 protein.

Type: Primary
Antigen: G3BP-1
Clonality: Polyclonal
Clone: D226
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR