Anti-GAD-67 Rabbit Polyclonal Antibody [clone: A492]

Lieferant: Bioworld Technology
BS1135
BWRLBS1135EA 262 EUR
BWRLBS1135
Anti-GAD-67 Rabbit Polyclonal Antibody [clone: A492]
Antikörper
Synthetic peptide, corresponding to amino acids 460-514 of Human GAD-67.

GAD67 (A492) pAb detects endogenous levels of GAD67 protein.

Type: Primary
Antigen: GAD-67
Clonality: Polyclonal
Clone: A492
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR