Anti-HDAC2 Rabbit Polyclonal Antibody [clone: D388]

Lieferant: Bioworld Technology
BS1162
BWRLBS1162EA 262 EUR
BWRLBS1162
Anti-HDAC2 Rabbit Polyclonal Antibody [clone: D388]
Antikörper
Synthetic peptide, corresponding to amino acids 356-400 of Human HDAC2.

HDAC2 (D388) pAb detects endogenous levels of HDAC2 protein.

Type: Primary
Antigen: HDAC2
Clonality: Polyclonal
Clone: D388
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR