Anti-HDAC8 Rabbit Polyclonal Antibody [clone: K33]
Lieferant: Bioworld Technology
BS1167
BWRLBS1167EA 262 EUR
BWRLBS1167
Anti-HDAC8 Rabbit Polyclonal Antibody [clone: K33]
Antikörper
Synthetic peptide, corresponding to amino acids 2-45 of Human HDAC8.
HDAC8 (K33) pAb detects endogenous levels of HDAC8 protein.
Type: Primary
Antigen: HDAC8
Clonality: Polyclonal
Clone: K33
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat