Anti-Ku-70 Rabbit Polyclonal Antibody [clone: R586]
Lieferant: Bioworld Technology
BS1212
BWRLBS1212EA 262 EUR
BWRLBS1212
Anti-Ku-70 Rabbit Polyclonal Antibody [clone: R586]
Antikörper
Synthetic peptide, corresponding to amino acids 560-600 of Human Ku-70.
Ku70 (R586) pAb detects endogenous levels of Ku70 protein.
Type: Primary
Antigen: Ku-70
Clonality: Polyclonal
Clone: R586
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat