Anti-Ku-80 Rabbit Polyclonal Antibody [clone: A717]
Lieferant: Bioworld Technology
BS1213
BWRLBS1213EA 262 EUR
BWRLBS1213
Anti-Ku-80 Rabbit Polyclonal Antibody [clone: A717]
Antikörper
Synthetic peptide, corresponding to amino acids 690-730 of Human Ku-80.
Ku80(A717) pAb detects endogenous levels of Ku80 protein.
Type: Primary
Antigen: Ku-80
Clonality: Polyclonal
Clone: A717
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat