Anti-MMP-19 Rabbit Polyclonal Antibody [clone: G44]

Lieferant: Bioworld Technology
BS1235
BWRLBS1235EA 262 EUR
BWRLBS1235
Anti-MMP-19 Rabbit Polyclonal Antibody [clone: G44]
Antikörper
Synthetic peptide, corresponding to amino acids 12-60 of Human MMP-19.

MMP-19 (G44) pAb detects endogenous levels of MMP-19 protein.

Type: Primary
Antigen: MMP-19
Clonality: Polyclonal
Clone: G44
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR