Anti-NFkB-p100 Rabbit Polyclonal Antibody [clone: S865]

Lieferant: Bioworld Technology
BS1246
BWRLBS1246EA 262 EUR
BWRLBS1246
Anti-NFkB-p100 Rabbit Polyclonal Antibody [clone: S865]
Antikörper
Synthetic peptide, corresponding to amino acids 836-886 of Human NFkB-p100.

NFkB-p100 (S865) pAb detects endogenous levels of NFkB-p100 protein.This antibody does not recognize NFkB-p52 subunit.

Type: Primary
Antigen: NFkB-p100
Clonality: Polyclonal
Clone: S865
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR