Anti-NFkB-p65 Rabbit Polyclonal Antibody [clone: M270]

Lieferant: Bioworld Technology
BS1253
BWRLBS1253EA 262 EUR
BWRLBS1253
Anti-NFkB-p65 Rabbit Polyclonal Antibody [clone: M270]
Antikörper
Synthetic peptide, corresponding to amino acids 250-300 of Human NFkB-p65.

NF?B-p65 (M270) pAb detects endogenous levels of NF?B-p65 protein.

Type: Primary
Antigen: NFkB-p65
Clonality: Polyclonal
Clone: M270
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR