Anti-p53 Rabbit Polyclonal Antibody [clone: P309]

Lieferant: Bioworld Technology
BS1274
BWRLBS1274EA 262 EUR
BWRLBS1274
Anti-p53 Rabbit Polyclonal Antibody [clone: P309]
Antikörper
Synthetic peptide, corresponding to amino acids 271-310 of Human p53.

p53 (P309) pAb detects endogenous levels of p53 protein.

Type: Primary
Antigen: p53
Clonality: Polyclonal
Clone: P309
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR