Anti-PR Rabbit Polyclonal Antibody [clone: V184]

Lieferant: Bioworld Technology
BS1299
BWRLBS1299EA 262 EUR
BWRLBS1299
Anti-PR Rabbit Polyclonal Antibody [clone: V184]
Antikörper
Synthetic peptide, corresponding to amino acids 151-200 of Human PR.

Progesterone Receptor (V184) pAb detects endogenous levels of Progesterone Receptor protein.

Type: Primary
Antigen: PR
Clonality: Polyclonal
Clone: V184
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR