Anti-PAR4 Rabbit Polyclonal Antibody [clone: D313]

Lieferant: Bioworld Technology
BS1300
BWRLBS1300EA 262 EUR
BWRLBS1300
Anti-PAR4 Rabbit Polyclonal Antibody [clone: D313]
Antikörper
Synthetic peptide, corresponding to amino acids 283-332 of Human PAR4.

Prostate Apoptosis Response protein-4 (D313) pAb detects endogenous levels of Prostate Apoptosis Response protein-4 protein.

Type: Primary
Antigen: PAR4
Clonality: Polyclonal
Clone: D313
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR