Anti-RCBTB1 Rabbit Polyclonal Antibody [clone: A277]
Lieferant: Bioworld Technology
BS1313
BWRLBS1313EA 262 EUR
BWRLBS1313
Anti-RCBTB1 Rabbit Polyclonal Antibody [clone: A277]
Antikörper
Synthetic peptide, corresponding to amino acids 245-290 of Human RCBTB1.
RCBTB1 (A277) pAb detects endogenous levels of RCBTB1 protein.
Type: Primary
Antigen: RCBTB1
Clonality: Polyclonal
Clone: A277
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse