Anti-c-Src Rabbit Polyclonal Antibody [clone: L412]

Lieferant: Bioworld Technology
BS1330
BWRLBS1330EA 262 EUR
BWRLBS1330
Anti-c-Src Rabbit Polyclonal Antibody [clone: L412]
Antikörper
Synthetic peptide, corresponding to amino acids 390-430 of Human c-Src.

Src (L412) pAb detects endogenous levels of Src protein.

Type: Primary
Antigen: c-Src
Clonality: Polyclonal
Clone: L412
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR