Anti-Stat3 Rabbit Polyclonal Antibody [clone: T721]

Lieferant: Bioworld Technology
BS1336
BWRLBS1336EA 262 EUR
BWRLBS1336
Anti-Stat3 Rabbit Polyclonal Antibody [clone: T721]
Antikörper
Synthetic peptide, corresponding to amino acids 700-750 of Human Stat3.

STAT3 (T721) pAb detects endogenous levels of STAT3 protein.

Type: Primary
Antigen: Stat3
Clonality: Polyclonal
Clone: T721
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR