Anti-TH Rabbit Polyclonal Antibody [clone: G13]

Lieferant: Bioworld Technology
BS1368
BWRLBS1368EA 262 EUR
BWRLBS1368
Anti-TH Rabbit Polyclonal Antibody [clone: G13]
Antikörper
Synthetic peptide, corresponding to the N-terminus of Human TH.

Tyrosine Hydroxylase (G13) pAb detects endogenous levels of Tyrosine Hydroxylase protein.

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone: G13
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR