Anti-4E-BP1 Rabbit Polyclonal Antibody [clone: L58]

Lieferant: Bioworld Technology
BS1375
BWRLBS1375EA 262 EUR
BWRLBS1375
Anti-4E-BP1 Rabbit Polyclonal Antibody [clone: L58]
Antikörper
Synthetic peptide, corresponding to amino acids 26-76 of Human 4E-BP1.

4E-BP1 (L58) pAb detects endogenous levels of 4E-BP1 protein.

Type: Primary
Antigen: 4E-BP1
Clonality: Polyclonal
Clone: L58
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR