Anti-C/EBP-alpha Rabbit Polyclonal Antibody [clone: M15]

Lieferant: Bioworld Technology
BS1384
BWRLBS1384EA 262 EUR
BWRLBS1384
Anti-C/EBP-alpha Rabbit Polyclonal Antibody [clone: M15]
Antikörper
Synthetic peptide, corresponding to amino acids N-terminus of Human C/EBP-alpha.

C/EBP-? (M15) pAb detects endogenous levels of C/EBP-? protein.

Type: Primary
Antigen: C/EBP-alpha
Clonality: Polyclonal
Clone: M15
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR