Anti-Chk2 Rabbit Polyclonal Antibody [clone: M381]

Lieferant: Bioworld Technology
BS1391
BWRLBS1391EA 262 EUR
BWRLBS1391
Anti-Chk2 Rabbit Polyclonal Antibody [clone: M381]
Antikörper
Synthetic peptide, corresponding to amino acids 351-400 of Human Chk2.

Chk2 (M381) pAb detects endogenous levels of Chk2 protein.

Type: Primary
Antigen: Chk2
Clonality: Polyclonal
Clone: M381
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR