Anti-Ku-80 Rabbit Polyclonal Antibody [clone: D708]
Lieferant: Bioworld Technology
BS1409
BWRLBS1409EA 262 EUR
BWRLBS1409
Anti-Ku-80 Rabbit Polyclonal Antibody [clone: D708]
Antikörper
Synthetic peptide, corresponding to amino acids 680-720 of Human Ku-80.
Ku80 (D708) pAb detects endogenous levels of Ku80 protein.
Type: Primary
Antigen: Ku-80
Clonality: Polyclonal
Clone: D708
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human