Anti-Trk A Rabbit Polyclonal Antibody [clone: A785]
Lieferant: Bioworld Technology
BS1430
BWRLBS1430EA 262 EUR
BWRLBS1430
Anti-Trk A Rabbit Polyclonal Antibody [clone: A785]
Antikörper
Synthetic peptide, corresponding to amino acids 751-800 of Human Trk A.
Trk A (A785) pAb detects endogenous levels of Trk A protein.
Type: Primary
Antigen: Trk A
Clonality: Polyclonal
Clone: A785
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human