Anti-Ku-86 Rabbit Polyclonal Antibody [clone: A717]

Lieferant: Bioworld Technology
BS1497
BWRLBS1497EA 262 EUR
BWRLBS1497
Anti-Ku-86 Rabbit Polyclonal Antibody [clone: A717]
Antikörper
Synthetic peptide, corresponding to amino acids 700-730 of Human Ku-86.

XRCC5 (A717) pAb detects endogenous levels of XRCC5 protein.

Type: Primary
Antigen: Ku-86
Clonality: Polyclonal
Clone: A717
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR