Anti-APC Rabbit Polyclonal Antibody [clone: K2050]

Lieferant: Bioworld Technology
BS1507
BWRLBS1507EA 262 EUR
BWRLBS1507
Anti-APC Rabbit Polyclonal Antibody [clone: K2050]
Antikörper
Synthetic peptide, corresponding to amino acids 2020-2070 of Human APC.

APC (K2050) pAb detects endogenous levels of APC protein.

Type: Primary
Antigen: APC
Clonality: Polyclonal
Clone: K2050
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR