Anti-Arrestin-beta-1 Rabbit Polyclonal Antibody [clone: E406]
Lieferant: Bioworld Technology
BS1509
BWRLBS1509EA 262 EUR
BWRLBS1509
Anti-Arrestin-beta-1 Rabbit Polyclonal Antibody [clone: E406]
Antikörper
Synthetic peptide, corresponding to amino acids 390-440 of Human Arrestin-beta-1.
Arrestin-?-1 (E406) pAb detects endogenous levels of Arrestin-?-1 protein.
Type: Primary
Antigen: Arrestin-beta-1
Clonality: Polyclonal
Clone: E406
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat