Anti-eIF4G Rabbit Polyclonal Antibody [clone: Q1103]

Lieferant: Bioworld Technology
BS1534
BWRLBS1534EA 262 EUR
BWRLBS1534
Anti-eIF4G Rabbit Polyclonal Antibody [clone: Q1103]
Antikörper
Synthetic peptide, corresponding to amino acids 1080-1130 of Human eIF4G.

eIF4G (Q1103) pAb detects endogenous levels of eIF4G protein.

Type: Primary
Antigen: eIF4G
Clonality: Polyclonal
Clone: Q1103
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR