Anti-MYPT1 Rabbit Polyclonal Antibody [clone: R847]

Lieferant: Bioworld Technology
BS1558
BWRLBS1558EA 262 EUR
BWRLBS1558
Anti-MYPT1 Rabbit Polyclonal Antibody [clone: R847]
Antikörper
Synthetic peptide, corresponding to amino acids 821-870 of Human MYPT1.

MYPT1 (R847) pAb detects endogenous levels of MYPT1 protein.

Type: Primary
Antigen: MYPT1
Clonality: Polyclonal
Clone: R847
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR